KEY465195275 Key Organics / BIONET

(3E)-1,1,1-trifluoro-4-{[2-(trifluoromethoxy)phenyl]amino}but-3-en-2-one

  • MDL number MFCD02186436
  • Cas Number : 1164558-76-2
  • Molecular Weight : 299.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72791

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0722_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart