KEY465195286 Key Organics / BIONET

(2E)-1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-3-(pyridin-3-yl)prop-2-en-1-one

  • MDL number MFCD28347909
  • Cas Number : 1823194-60-0
  • Molecular Weight : 340.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 212
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453055

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart