KEY465195287 Key Organics / BIONET

5-methyl-4-(naphthalene-2-carbonyl)-1-(4-nitrophenyl)-1H-1,2,3-triazole

  • MDL number MFCD02571924
  • Cas Number : 478041-07-5
  • Molecular Weight : 358.357

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72808

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart