KEY465195300 Key Organics / BIONET

1-[4-(4-nitro-1H-1,2,3-triazol-1-yl)phenyl]ethan-1-one

  • MDL number MFCD02186228
  • Cas Number : 478041-21-3
  • Molecular Weight : 232.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72827

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0851_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart