KEY465195303 Key Organics / BIONET

(2E)-3-[(4-methoxyphenyl)amino]-1-[4-methyl-2-(pyridin-3-yl)-1,3-thiazol-5-yl]prop-2-en-1-one

  • MDL number MFCD02186230
  • Cas Number : 478041-29-1
  • Molecular Weight : 351.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72835

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0861_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart