KEY465195305 Key Organics / BIONET

methyl (2Z)-3-[(4-chloro-2-fluorophenyl)amino]-2-phenylbut-2-enoate

  • MDL number MFCD02186451
  • Cas Number : 478041-31-5
  • Molecular Weight : 319.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72837

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0864_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart