KEY465195311 Key Organics / BIONET

1-{4-[4-(2,4-dichlorobenzoyl)-5-methyl-1H-1,2,3-triazol-1-yl]phenyl}ethan-1-one

  • MDL number MFCD02571932
  • Cas Number : 478041-40-6
  • Molecular Weight : 374.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72847

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0878_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart