KEY465195319 Key Organics / BIONET

4-[1-(benzenesulfonyl)piperidine-4-carbonyl]-2,6-dimethylmorpholine

  • MDL number MFCD02069180
  • Cas Number : 326871-46-9
  • Molecular Weight : 366.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72865

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-1021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart