KEY465195333 Key Organics / BIONET

1-(4-chlorobenzenesulfonyl)-4-[(morpholin-4-yl)methyl]piperidin-4-ol

  • MDL number MFCD02186638
  • Cas Number : 478041-61-1
  • Molecular Weight : 374.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72896

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-1069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart