KEY465195342 Key Organics / BIONET

(1E)-1-(4-methylphenyl)-N-{1H-pyrrolo[2,3-b]pyridin-1-yl}methanimine

  • MDL number MFCD03102809
  • Cas Number : 861212-75-1
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72913

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1090_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart