KEY465195344 Key Organics / BIONET

(2E)-3-(2,4-dichlorophenyl)-N'-(4-methylbenzenesulfonyl)prop-2-enehydrazide

  • MDL number MFCD02187061
  • Cas Number : 478041-75-7
  • Molecular Weight : 385.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72916

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-1093_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart