KEY465195350 Key Organics / BIONET

1-(2-{[(4-chlorophenyl)methyl]sulfanyl}-4-methylpyrimidin-5-yl)ethan-1-one

  • MDL number MFCD02187392
  • Cas Number : 478041-93-9
  • Molecular Weight : 292.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72948

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-1174_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart