KEY465195351 Key Organics / BIONET

1-(2-{[(3,4-dichlorophenyl)methyl]sulfanyl}-4-methylpyrimidin-5-yl)ethan-1-one

  • MDL number MFCD02187393
  • Cas Number : 478041-94-0
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72949

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1175_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart