KEY465195356 Key Organics / BIONET

1-{6-chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one

  • MDL number MFCD00973744
  • Cas Number : 154877-60-8
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72956

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-1184_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart