KEY465195359 Key Organics / BIONET

1-{2,7-dimethylimidazo[1,2-a]pyridin-3-yl}-2-[(4-methylphenyl)sulfanyl]ethan-1-one

  • MDL number MFCD02186771
  • Cas Number : 478042-00-1
  • Molecular Weight : 310.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72961

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1194_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart