KEY465195360 Key Organics / BIONET

2-[(3-chlorophenyl)sulfanyl]-1-{2,7-dimethylimidazo[1,2-a]pyridin-3-yl}ethan-1-one

  • MDL number MFCD02186772
  • Cas Number : 478042-01-2
  • Molecular Weight : 330.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72962

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1195_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart