KEY465195370 Key Organics / BIONET

(3Z)-2-(propan-2-ylsulfanyl)-3-[2-(2,4,6-trichlorophenyl)hydrazin-1-ylidene]-3H-indole

  • MDL number MFCD02187071
  • Cas Number : 478042-26-1
  • Molecular Weight : 398.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72989

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-1264_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart