KEY465195376 Key Organics / BIONET

1-phenyl-2-(1H-1,2,4-triazol-3-ylsulfanyl)ethan-1-one

  • MDL number MFCD03012276
  • Cas Number : 32189-00-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73053

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart