KEY465195379 Key Organics / BIONET

ethyl (2Z)-4,4,4-trifluoro-3-hydroxy-2-(methoxyimino)butanoate

  • MDL number MFCD03012126
  • Cas Number : 102608-31-1
  • Molecular Weight : 229.155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73068

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0051_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart