KEY465195382 Key Organics / BIONET

4-{4-[(3,4-dimethoxyphenyl)methyl]-5-methyl-4H-1,2,4-triazol-3-yl}-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD03305642
  • Cas Number : 956371-02-1
  • Molecular Weight : 328.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73090

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0217_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart