KEY465195384 Key Organics / BIONET

methyl 2-[(4-cyano-1-phenyl-5,6,7,8-tetrahydroisoquinolin-3-yl)sulfanyl]acetate

  • MDL number MFCD03305645
  • Cas Number : 861212-86-4
  • Molecular Weight : 338.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73094

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0221_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart