KEY465195389 Key Organics / BIONET

1-amino-5-phenyl-6H,7H,8H,9H-thieno[2,3-c]isoquinoline-2-carbonitrile

  • MDL number MFCD00582242
  • Cas Number : 296878-08-5
  • Molecular Weight : 305.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73108

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0249_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart