KEY465195390 Key Organics / BIONET

5-{[6-(4-methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methylidene}-1,3-diazinane-2,4,6-trione

  • MDL number MFCD03305655
  • Cas Number : 861212-97-7
  • Molecular Weight : 352.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0283_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart