KEY465195394 Key Organics / BIONET

2-{[6-(4-methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]methylidene}-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD03305659
  • Cas Number : 861213-01-6
  • Molecular Weight : 370.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73130

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart