KEY465195395 Key Organics / BIONET

1,3-dimethyl-5-{[4-(1H-1,2,4-triazol-1-yl)phenyl]methylidene}-1,3-diazinane-2,4,6-trione

  • MDL number MFCD03012139
  • Cas Number : 439107-33-2
  • Molecular Weight : 311.301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73135

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0304_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart