KEY465195398 Key Organics / BIONET

methyl 2-[4-(naphthalene-2-sulfonyl)-3,4-dihydro-2H-1,4-benzoxazin-3-yl]acetate

  • MDL number MFCD03787319
  • Cas Number : 865657-69-8
  • Molecular Weight : 397.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73144

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0317_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart