KEY465195400 Key Organics / BIONET

3-[(cyanomethyl)sulfanyl]-1-(3-methylphenyl)-5,6,7,8-tetrahydroisoquinoline-4-carbonitrile

  • MDL number MFCD03012143
  • Cas Number : 439107-38-7
  • Molecular Weight : 319.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73150

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0325_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart