KEY465195406 Key Organics / BIONET

N-[2-amino-4-(4-methylpiperazin-1-yl)phenyl]-3-chlorobenzamide

  • MDL number MFCD03012539
  • Cas Number : 439107-46-7
  • Molecular Weight : 344.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73169

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0349_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart