KEY465195408 Key Organics / BIONET

(2E)-1-(3-bromothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one

  • MDL number MFCD03618051
  • Cas Number : 865657-77-8
  • Molecular Weight : 274.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73182

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0604_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart