KEY465195411 Key Organics / BIONET

N-[(2Z)-5,6-dimethoxy-3-oxo-2-[(thiophen-2-yl)methylidene]-2,3-dihydro-1H-inden-1-yl]-2,2,2-trifluoroacetamide

  • MDL number MFCD03848444
  • Cas Number : 865657-79-0
  • Molecular Weight : 397.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0640_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart