KEY465195412 Key Organics / BIONET

6-iodo-2-(3-nitrophenyl)imidazo[1,2-a]pyridine

  • MDL number MFCD00438340
  • Cas Number : 865657-80-3
  • Molecular Weight : 365.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73206

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0653_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart