KEY465195413 Key Organics / BIONET

3-(2-bromophenyl)-3-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD04125168
  • Cas Number : 117291-11-9
  • Molecular Weight : 340.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73208

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0659_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart