KEY465195415 Key Organics / BIONET

N-(5-acetyl-4-methyl-1,3-thiazol-2-yl)-N-phenylbenzamide

  • MDL number MFCD27924212
  • Cas Number : 1797764-64-7
  • Molecular Weight : 336.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135 - 137
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402883

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0807_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart