KEY465195417 Key Organics / BIONET

4-(4-methoxyphenyl)-1-(4-phenyl-1,3-thiazol-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD03305670
  • Cas Number : 956961-86-7
  • Molecular Weight : 348.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73218

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart