KEY465195418 Key Organics / BIONET

1-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-4-(4-methoxyphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD03012166
  • Cas Number : 956961-87-8
  • Molecular Weight : 382.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart