KEY465195420 Key Organics / BIONET

methyl (3E)-3-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazin-1-ylidene}-2-(1H-1,2,4-triazol-1-yl)propanoate

  • MDL number MFCD03012169
  • Cas Number : 551930-85-9
  • Molecular Weight : 362.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73223

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart