KEY465195427 Key Organics / BIONET

1-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]-4-(thiophen-3-yl)-1H-pyrazol-5-amine

  • MDL number MFCD03305674
  • Cas Number : 955967-07-4
  • Molecular Weight : 354.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73233

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0840_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart