KEY465195433 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-[2-(4-fluorophenyl)-4-methyl-1,3-thiazol-5-yl]prop-2-en-1-one

  • MDL number MFCD27924213
  • Cas Number : 1382748-03-9
  • Molecular Weight : 290.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207 - 209
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402884

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0881_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart