KEY465195435 Key Organics / BIONET

3-[4-(3-chlorophenyl)piperazin-1-yl]-1H-1,2,4-triazol-5-amine

  • MDL number MFCD03305688
  • Cas Number : 865657-91-6
  • Molecular Weight : 278.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73262

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0887_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart