KEY465195450 Key Organics / BIONET

5-(3-chlorophenoxy)-1,3-diphenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD03012908
  • Cas Number : 956368-13-1
  • Molecular Weight : 374.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-1123_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart