KEY465195452 Key Organics / BIONET

(4,5-dibromo-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate

  • MDL number MFCD03012927
  • Cas Number : 231613-44-8
  • Molecular Weight : 325.944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73337

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-1493_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart