KEY465195453 Key Organics / BIONET

2-methyl-6-(4-methylphenyl)-4-(3,4,5-trimethoxyphenyl)pyridine-3-carbonitrile

  • MDL number MFCD03848446
  • Cas Number : 865658-16-8
  • Molecular Weight : 374.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart