KEY465195458 Key Organics / BIONET

[3-(3,4-dimethoxyphenyl)-1-phenyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD02928876
  • Cas Number : 429620-48-4
  • Molecular Weight : 310.353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73355

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0202_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart