KEY465195461 Key Organics / BIONET

[3-(naphthalen-2-yl)-1-phenyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD02720236
  • Cas Number : 36640-67-2
  • Molecular Weight : 300.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73360

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart