KEY465195465 Key Organics / BIONET

4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-(quinolin-6-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04125181
  • Cas Number : 865658-27-1
  • Molecular Weight : 362.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73370

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0229_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart