KEY465195466 Key Organics / BIONET

2-[1-acetyl-2-(3,4-dichlorophenyl)-5-methyl-3-oxo-2,3-dihydro-1H-pyrazol-4-yl]ethyl acetate

  • MDL number MFCD03787074
  • Cas Number : 865658-32-8
  • Molecular Weight : 371.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0241_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart