KEY465195472 Key Organics / BIONET

2-(5-methyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)ethyl 4-methylbenzoate

  • MDL number MFCD03848464
  • Cas Number : 865658-46-4
  • Molecular Weight : 336.391

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73404

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0308_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart