KEY465195473 Key Organics / BIONET

N-(6-acetyl-2H-1,3-benzodioxol-5-yl)-3-chlorobenzamide

  • MDL number MFCD03787182
  • Cas Number : 698985-86-3
  • Molecular Weight : 317.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73420

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0341_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart