KEY465195479 Key Organics / BIONET

4-(4-chlorophenyl)-3,7,7-trimethyl-2H,4H,5H,6H,7H,8H,9H-pyrazolo[3,4-b]quinolin-5-one

  • MDL number MFCD03787089
  • Cas Number : 370588-24-2
  • Molecular Weight : 341.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73441

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0443_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart