KEY465195480 Key Organics / BIONET

3,7,7-trimethyl-4-phenyl-2H,4H,5H,6H,7H,8H,9H-pyrazolo[3,4-b]quinolin-5-one

  • MDL number MFCD03787090
  • Cas Number : 329978-12-3
  • Molecular Weight : 307.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73442

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0444_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart